siRNA Details
| Gene Symbol: MYL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199629.1 | NP_001186558.1 | 6, 7 | 927 | ||
| NM_002475.4 | NP_002466.1 | 6, 7 | 863 | |||
| GenBank mRNA | AI349553.1 | - | 2, 3 | 142 | ||
| BC012425.1 | - | 5, 6 | 651 | 3862144 | ||
| BC014400.1 | - | 6, 7 | 927 | 3503030 | ||
| M31211.1 | - | 5, 6 | 631 | |||
| X16434.1 | - | 5, 6 | 616 | |||

MYL6B
Genomic Map