siRNA Details
| Gene Symbol: MROH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152503.5 | NP_689716.4 | 7 | 1030 | ||
| NM_213631.2 | NP_998796.1 | 7 | 1030 | |||
| NM_213632.2 | NP_998797.2 | 7 | 1030 | |||
| GenBank mRNA | AK093432.1 | - | 7 | 1030 | ||
| AL833864.1 | - | 1 | 32 | |||
| BC030006.1 | - | 7 | 1007 | 4822338 | ||
| BC057767.1 | - | 7 | 1013 | 5272440 | ||

MROH8
Genomic Map