siRNA Details
| Gene Symbol: SMCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144775.2 | NP_658988.2 | 1 | 1487 | ||
| GenBank mRNA | AF467440.1 | - | 1 | 1365 | ||
| AK296847.1 | - | 1 | 1475 | |||
| BC101115.1 | - | 1 | 1361 | |||
| BC101116.2 | - | 1 | 1361 | |||
| BC101117.2 | - | 1 | 1361 | |||
| BC142680.1 | - | 1 | 1457 | 40148405 | ||

SMCR8
Genomic Map