siRNA Details
| Gene Symbol: RBBP8NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080833.2 | NP_543023.2 | 5, 6 | 506 | ||
| GenBank mRNA | AK074919.1 | - | 5, 6 | 466 | ||
| AK290299.1 | - | 5, 6 | 479 | |||
| BC137326.1 | - | 4, 5 | 394 | 9021324 | ||
| BC137327.1 | - | 4, 5 | 394 | 9021325 | ||
| DA871775.1 | - | 5, 6 | 508 | |||

RBBP8NL
Genomic Map