siRNA Details
Gene Symbol: CNBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207076.2 | NP_001194005.1 | 7 | 966 | ||
NM_080834.3 | NP_543024.2 | 7 | 966 | |||
XM_005260295.2 | XP_005260352.1 | 7 | 940 | |||
XM_011528590.2 | XP_011526892.1 | 7 | 926 | |||
XM_011528592.1 | XP_011526894.1 | 6 | 796 | |||
XM_011528593.2 | XP_011526895.1 | 6 | 659 | |||
XM_011528598.1 | XP_011526900.1 | 3 | 296 | |||
XM_017027683.1 | XP_016883172.1 | 7 | 1144 | |||
GenBank mRNA | AK057454.1 | - | 7 | 940 | ||
BC114945.1 | - | 7 | 924 | 40084218 |
