siRNA Details
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080878.2 | NP_543154.1 | 8 | 967 | ||
| XM_011509210.1 | XP_011507512.1 | 10 | 1444 | |||
| XM_011509211.1 | XP_011507513.1 | 10 | 1441 | |||
| GenBank mRNA | AY065973.1 | - | 8 | 939 | ||
| AY358905.1 | - | 8 | 967 | |||
| BC117225.1 | - | 8 | 965 | 40125776 | ||
| BC143339.1 | - | 7 | 979 | |||
| BC143341.1 | - | 8 | 965 | 9051846 | ||

ITLN2
Genomic Map