siRNA Details
| Gene Symbol: ACSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207321.2 | NP_997204.2 | 6 | 1022 | ||
| GenBank mRNA | AK126134.1 | - | 6 | 1012 | ||
| BC063654.1 | - | 3 | 411 | |||
| BC104210.1 | - | 5 | 906 | |||
| BC104211.2 | - | 5 | 906 | |||
| BC112988.1 | - | 6 | 987 | |||
| BC148184.1 | - | 5 | 906 | |||

ACSM6
Genomic Map