siRNA Details
| Gene Symbol: SYT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175733.3 | NP_783860.1 | 5 | 1533 | ||
| XM_011519900.2 | XP_011518202.1 | 5 | 1511 | |||
| XM_011519901.2 | XP_011518203.1 | 5 | 1511 | |||
| XM_011519902.2 | XP_011518204.1 | 5 | 1284 | |||
| XM_011519904.2 | XP_011518206.1 | 5 | 1511 | |||
| GenBank mRNA | AK055003.1 | - | 4 | 766 | ||
| AK315013.1 | - | 5 | 1476 | |||
| BC029605.1 | - | 6 | 1443 | 5296925 | ||
| BC046367.1 | - | 5 | 1533 | 5770087 | ||

SYT9
Genomic Map