siRNA Details
| Gene Symbol: OR52B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005161.3 | NP_001005161.2 | 1 | 709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52B4| Gene Symbol: OR52B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005161.3 | NP_001005161.2 | 1 | 709 | ||