siRNA Details
| Gene Symbol: UBQLNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145053.4 | NP_659490.4 | 1 | 343 | ||
| GenBank mRNA | AK057255.1 | - | 1 | 346 | ||
| BC012183.1 | - | 1 | 203 | 4554912 | ||
| BC034977.2 | - | 1 | 312 | 4822902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBQLNL| Gene Symbol: UBQLNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145053.4 | NP_659490.4 | 1 | 343 | ||
| GenBank mRNA | AK057255.1 | - | 1 | 346 | ||
| BC012183.1 | - | 1 | 203 | 4554912 | ||
| BC034977.2 | - | 1 | 312 | 4822902 | ||