siRNA Details
| Gene Symbol: MUC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135091.1 | NP_001128563.1 | 5 | 1253 | ||
| NM_001135092.1 | NP_001128564.1 | 4 | 1103 | |||
| NM_145650.3 | NP_663625.2 | 4 | 1165 | |||
| XM_017017235.1 | XP_016872724.1 | 5 | 1253 | |||
| XM_017017236.1 | XP_016872725.1 | 4 | 1103 | |||
| GenBank mRNA | AJ417818.1 | - | 5 | 1121 | ||
| AJ507429.1 | - | 4 | 977 | |||
| AK026790.1 | - | 2 | 149 | |||
| AK128337.1 | - | 4 | 1018 | |||
| AY358668.1 | - | 5 | 1218 | |||
| BC020912.1 | - | 5 | 1245 | 4733074 | ||
| BC058007.1 | - | 5 | 1131 | 4719841 | ||

MUC15
Genomic Map