siRNA Details
| Gene Symbol: KBTBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152433.3 | NP_689646.2 | 3 | 2199 | ||
| NM_198439.2 | NP_940841.1 | 4 | 2400 | |||
| XM_006718767.3 | XP_006718830.1 | 4 | 2430 | |||
| XM_011542618.2 | XP_011540920.1 | 3 | 2044 | |||
| XM_011542619.2 | XP_011540921.1 | 3 | 2182 | |||
| GenBank mRNA | AK055247.1 | - | 1 | 816 | ||
| AK092993.1 | - | 3 | 1802 | |||
| AK289915.1 | - | 4 | 2010 | |||
| BC117466.1 | - | 3 | 1898 | 40126017 | ||
| BX640672.1 | - | 3 | 1920 | |||
| BX647830.1 | - | 3 | 1347 | |||

KBTBD3
Genomic Map