siRNA Details
| Gene Symbol: KDELC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153705.4 | NP_714916.3 | 6, 7 | 1348 | ||
| XM_011542621.2 | XP_011540923.1 | 6, 7 | 1321 | |||
| XM_011542623.2 | XP_011540925.1 | 4, 5 | 901 | |||
| GenBank mRNA | AF533708.1 | - | 6, 7 | 1307 | ||
| AK098206.1 | - | 2, 3 | 313 | |||
| AK313534.1 | - | 6, 7 | 1210 | |||
| AY358616.1 | - | 5, 6 | 1417 | |||
| BC036526.2 | - | 6, 7 | 1302 | 5266665 | ||

KDELC2
Genomic Map