siRNA Details
| Gene Symbol: AMDHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152435.2 | NP_689648.2 | 4, 5 | 685 | ||
| GenBank mRNA | AB075878.1 | - | 4, 5 | 640 | ||
| AK054617.1 | - | 4, 5 | 685 | |||
| AK122802.1 | - | 4, 5 | 652 | |||
| AK290828.1 | - | 4, 5 | 640 | |||
| AL833144.1 | - | 3, 4 | 393 | |||
| BC029146.1 | - | 4, 5 | 621 | 5183113 | ||

AMDHD1
Genomic Map