siRNA Details
| Gene Symbol: WDR66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178003.1 | NP_001171474.1 | 3 | 631 | ||
| NM_144668.5 | NP_653269.3 | 3 | 618 | |||
| GenBank mRNA | AK097102.1 | - | 2 | 372 | ||
| BC028421.2 | - | 3 | 616 | 4826893 | ||
| BC036233.1 | - | 3 | 611 | 5295406 | ||

WDR66
Genomic Map