siRNA Details
| Gene Symbol: SLC38A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172702.1 | NP_001166173.1 | 12, 13 | 1098 | ||
| NM_153811.2 | NP_722518.2 | 12, 13 | 1098 | |||
| NR_033344.1 | 12, 13 | 1274 | ||||
| XM_006720050.1 | XP_006720113.1 | 8, 9 | 541 | |||
| XM_017021020.1 | XP_016876509.1 | 12, 13 | 1050 | |||
| XM_017021021.1 | XP_016876510.1 | 11, 12 | 996 | |||
| XM_017021022.1 | XP_016876511.1 | 12, 13 | 1363 | |||
| XM_017021023.1 | XP_016876512.1 | 12, 13 | 1050 | |||
| XM_017021024.1 | XP_016876513.1 | 10, 11 | 930 | |||
| XM_017021025.1 | XP_016876514.1 | 12, 13 | 1341 | |||
| XM_017021026.1 | XP_016876515.1 | 12, 13 | 1355 | |||
| XM_017021027.1 | XP_016876516.1 | 10, 11 | 719 | |||
| XM_017021028.1 | XP_016876517.1 | 8, 9 | 541 | |||
| GenBank mRNA | AF543422.1 | - | 12, 13 | 1050 | ||
| AK303839.1 | - | 12, 13 | 1077 | |||
| BC110378.1 | - | 12, 13 | 959 | 5737780 | ||
| BX248072.1 | - | 12, 13 | 919 | |||
| BX648332.1 | - | 12, 13 | 972 | |||

SLC38A6
Genomic Map