siRNA Details
| Gene Symbol: TERB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152448.2 | NP_689661.1 | 6 | 512 | ||
| GenBank mRNA | BC029537.1 | - | 6 | 512 | 5298504 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TERB2| Gene Symbol: TERB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152448.2 | NP_689661.1 | 6 | 512 | ||
| GenBank mRNA | BC029537.1 | - | 6 | 512 | 5298504 | |