siRNA Details
| Gene Symbol: SPATA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304804.1 | NP_001291733.1 | 3 | 300 | ||
| NM_001304805.1 | NP_001291734.1 | 4 | 449 | |||
| NM_001304806.1 | NP_001291735.1 | 3 | 363 | |||
| NM_173499.4 | NP_775770.1 | 3 | 518 | |||
| XM_011521265.2 | XP_011519567.1 | 3 | 565 | |||
| GenBank mRNA | AY489187.1 | - | 3 | 482 | ||
| BC033979.2 | - | 3 | 455 | 5295377 | ||
| BC066292.1 | - | 3 | 477 | 5271997 | ||
| BC112390.1 | - | 2 | 846 | 5168326 | ||
| HY024478.1 | - | 3 | 326 | |||
| HY047940.1 | - | 3 | 300 | |||

SPATA8
Genomic Map