siRNA Details
| Gene Symbol: ZNF688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024683.1 | NP_001019854.1 | 3 | 756 | ||
| NM_145271.3 | NP_660314.1 | 3 | 1444 | |||
| XM_005255139.2 | XP_005255196.1 | 3 | 673 | |||
| XM_005255140.2 | XP_005255197.1 | 3 | 1293 | |||
| GenBank mRNA | AF085979.1 | - | 1 | 96 | ||
| AK122680.1 | - | 3 | 1772 | |||
| BC018997.2 | - | 3 | 723 | 4336167 | ||
| KU178870.1 | - | 3 | 666 | |||
| KU178871.1 | - | 2 | 976 | |||
| KU178872.1 | - | 2 | 552 | |||

ZNF688
Genomic Map