siRNA Details
| Gene Symbol: TMEM199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152464.2 | NP_689677.1 | 5 | 588 | ||
| GenBank mRNA | AK223584.1 | - | 5 | 522 | ||
| AK291475.1 | - | 5 | 511 | |||
| AK311363.1 | - | 470 | ||||
| AY074907.1 | - | 5 | 498 | |||
| BC033113.1 | - | 5 | 505 | 5429060 | ||

TMEM199
Genomic Map