siRNA Details
| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144683.3 | NP_653284.2 | 4 | 664 | ||
| GenBank mRNA | AK296017.1 | - | 2 | 315 | ||
| AY358566.1 | - | 4 | 664 | |||
| BC015582.2 | - | 3 | 654 | 4637504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHRS13| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144683.3 | NP_653284.2 | 4 | 664 | ||
| GenBank mRNA | AK296017.1 | - | 2 | 315 | ||
| AY358566.1 | - | 4 | 664 | |||
| BC015582.2 | - | 3 | 654 | 4637504 | ||