siRNA Details
| Gene Symbol: TMEM99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195386.1 | NP_001182315.1 | 3 | 707 | ||
| NM_001195387.1 | NP_001182316.1 | 3 | 691 | |||
| NM_145274.3 | NP_660317.2 | 3 | 699 | |||
| XM_005257089.4 | XP_005257146.1 | 4 | 896 | |||
| XM_017024253.1 | XP_016879742.1 | 4 | 864 | |||
| GenBank mRNA | AI301318.1 | - | 1 | 178 | ||
| AK097454.1 | - | 3 | 640 | |||
| AK298616.1 | - | 3 | 642 | |||
| BC015365.1 | - | 3 | 678 | 3900532 | ||

TMEM99
Genomic Map