siRNA Details
Gene Symbol: AQP4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026908.1 | 6 | 481 | |||
GenBank mRNA | AK055069.1 | - | 6 | 481 | ||
BC105127.1 | - | 2 | 150 | |||
BC112070.1 | - | 2 | 150 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AQP4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026908.1 | 6 | 481 | |||
GenBank mRNA | AK055069.1 | - | 6 | 481 | ||
BC105127.1 | - | 2 | 150 | |||
BC112070.1 | - | 2 | 150 |