siRNA Details
| Gene Symbol: ANKRD29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308238.1 | NP_001295167.1 | 3, 4 | 392 | ||
| NM_173505.3 | NP_775776.2 | 3, 4 | 392 | |||
| XM_011525831.2 | XP_011524133.1 | 2, 3 | 2332 | |||
| XM_011525832.2 | XP_011524134.1 | 2, 3 | 2450 | |||
| GenBank mRNA | AK057782.1 | - | 3, 4 | 322 | ||
| AK313667.1 | - | 3, 4 | 310 | |||
| BC030622.1 | - | 3, 4 | 397 | 4818619 | ||

ANKRD29
Genomic Map