siRNA Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297624.1 | NP_001284553.1 | 4 | 602 | ||
| NM_001297625.1 | NP_001284554.1 | 4 | 604 | |||
| NM_144684.3 | NP_653285.2 | 5 | 731 | |||
| XM_011526465.2 | XP_011524767.1 | 5 | 1133 | |||
| GenBank mRNA | AK096442.1 | - | 3 | 467 | ||
| AK096559.1 | - | 5 | 721 | |||
| AK303271.1 | - | 4 | 593 | |||
| AL832560.1 | - | 4 | 581 | |||
| AY512662.1 | - | 5 | 700 | |||
| BC018783.1 | - | 5 | 700 | 4873910 | ||
| BC065503.1 | - | 5 | 706 | |||

ZNF480
Genomic Map