siRNA Details
| Gene Symbol: HIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144685.4 | NP_653286.2 | 3 | 1168 | ||
| XM_006723036.2 | XP_006723099.1 | 3 | 1143 | |||
| GenBank mRNA | AK292228.1 | - | 3 | 1129 | ||
| AK301713.1 | - | 4 | 921 | |||
| AK302301.1 | - | 3 | 525 | |||
| BC034501.2 | - | 3 | 1134 | 4823304 | ||

HIPK4
Genomic Map