siRNA Details
| Gene Symbol: TMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145303.2 | NP_001138775.2 | 12, 13 | 1969 | ||
| NM_144686.3 | NP_653287.2 | 12, 13 | 1951 | |||
| XM_011526486.2 | XP_011524788.1 | 9, 10 | 1454 | |||
| GenBank mRNA | AK314973.1 | - | 12, 13 | 1872 | ||
| AY236492.1 | - | 12, 13 | 1968 | |||
| AY263166.1 | - | 12, 13 | 1848 | |||
| BC025323.1 | - | 12, 13 | 1858 | 2959382 | ||
| BC032477.1 | - | 6, 7 | 1059 | |||
| BU680152.1 | - | 1, 2 | 80 | |||

TMC4
Genomic Map