siRNA Details
| Gene Symbol: ZNF542P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003127.2 | 7 | 1514 | |||
| NR_024055.2 | 6 | 1417 | ||||
| NR_024056.2 | 6 | 1670 | ||||
| NR_024057.2 | 5 | 1598 | ||||
| NR_033418.1 | 6 | 1738 | ||||
| GenBank mRNA | AK097128.1 | - | 5 | 1402 | ||
| AL833621.1 | - | 5 | 1367 | |||
| BC089419.1 | - | 5 | 1342 | |||
| BX640680.1 | - | 7 | 1474 | |||
| BX647919.1 | - | 6 | 1473 | |||

ZNF542P
Genomic Map