siRNA Details
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 9 | 1131 | ||
NM_207325.2 | NP_997208.2 | 9 | 1157 | |||
XM_006723045.1 | XP_006723108.1 | 6 | 673 | |||
XM_011526526.2 | XP_011524828.1 | 9 | 1112 | |||
XM_017026366.1 | XP_016881855.1 | 9 | 1148 | |||
XM_017026367.1 | XP_016881856.1 | 9 | 1071 | |||
GenBank mRNA | AK126264.1 | - | 3 | 232 | ||
AK126757.1 | - | 9 | 1148 | |||
AK129497.1 | - | 9 | 1112 | |||
BC131725.1 | - | 8 | 979 | 40123868 | ||
CR749459.1 | - | 3 | 327 |
