siRNA Details
| Gene Symbol: ZNF599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007248.2 | NP_001007249.1 | 3 | 566 | ||
| XM_017026369.1 | XP_016881858.1 | 3 | 641 | |||
| GenBank mRNA | AK055225.1 | - | 3 | 552 | ||
| AK302548.1 | - | 4 | 666 | |||
| AK307339.1 | - | 3 | 530 | |||
| BC044615.1 | - | 4 | 682 | 5266705 | ||
| BC092428.1 | - | 3 | 566 | |||

ZNF599
Genomic Map