siRNA Details
| Gene Symbol: FAM187B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152481.1 | NP_689694.1 | 1 | 327 | ||
| GenBank mRNA | AK098526.1 | - | 1 | 327 | ||
| BC039080.1 | - | 1 | 278 | |||
| BC117457.1 | - | 1 | 331 | 40126008 | ||
| BC117459.1 | - | 1 | 331 | 40126010 | ||

FAM187B
Genomic Map