siRNA Details
| Gene Symbol: ATP8B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178002.2 | NP_001171473.1 | 27 | 3595 | ||
| NM_138813.3 | NP_620168.1 | 27 | 3686 | |||
| NR_047593.2 | 27 | 3830 | ||||
| XM_006722654.2 | XP_006722717.1 | 27 | 3716 | |||
| XM_006722655.2 | XP_006722718.1 | 27 | 3795 | |||
| XM_006722656.2 | XP_006722719.1 | 27 | 3604 | |||
| XM_011527707.1 | XP_011526009.1 | 27 | 3689 | |||
| XM_011527708.1 | XP_011526010.1 | 27 | 3596 | |||
| XM_011527709.1 | XP_011526011.1 | 26 | 3503 | |||
| XM_011527710.2 | XP_011526012.1 | 21 | 2925 | |||
| XM_011527712.1 | XP_011526014.1 | 12 | 1718 | |||
| GenBank mRNA | AK057452.1 | - | 4 | 546 | ||
| AK125969.1 | - | 27 | 3830 | |||
| AY302538.1 | - | 27 | 3558 | |||
| BC035162.2 | - | 27 | 3681 | 5265541 | ||

ATP8B3
Genomic Map