siRNA Details
| Gene Symbol: ZNF555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172775.1 | NP_001166246.1 | 4 | 1785 | ||
| NM_152791.4 | NP_690004.4 | 4 | 1788 | |||
| XM_011527716.2 | XP_011526018.1 | 4 | 1788 | |||
| XM_017026375.1 | XP_016881864.1 | 4 | 1788 | |||
| GenBank mRNA | AK056659.1 | - | 1 | 1086 | ||
| AK093163.1 | - | 6 | 1521 | |||
| AK292954.1 | - | 4 | 1733 | |||
| AL832140.1 | - | 4 | 1749 | |||
| BC022022.2 | - | 4 | 1739 | 4822197 | ||
| BX647423.1 | - | 4 | 1746 | |||

ZNF555
Genomic Map