siRNA Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153812.2 | NP_722519.2 | 3 | 1010 | ||
| XM_011540762.1 | XP_011539064.1 | 3 | 812 | |||
| GenBank mRNA | AB209544.1 | - | 3 | 702 | ||
| AK027492.1 | - | 2 | 535 | |||
| AK097715.1 | - | 3 | 944 | |||
| AK315110.1 | - | 3 | 942 | |||
| AL121733.1 | - | 1 | 498 | |||
| AY069975.1 | - | 3 | 640 | |||
| BC032792.2 | - | 1 | 437 | |||
| BC038516.1 | - | 3 | 1021 | 5266750 | ||

PHF13
Genomic Map