siRNA Details
| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152489.1 | NP_689702.1 | 5 | 634 | ||
| XM_005270498.4 | XP_005270555.1 | 5 | 1428 | |||
| XM_006710379.3 | XP_006710442.1 | 5 | 569 | |||
| XM_011540764.2 | XP_011539066.1 | 6 | 1369 | |||
| XM_017000379.1 | XP_016855868.1 | 1 | 22 | |||
| XM_017000380.1 | XP_016855869.1 | 1 | 22 | |||
| XM_017000381.1 | XP_016855870.1 | 1 | 20 | |||
| XM_017000383.1 | XP_016855872.1 | 1 | 22 | |||
| XM_017000385.1 | XP_016855874.1 | 1 | 22 | |||
| GenBank mRNA | BC029895.1 | - | 5 | 634 | 5168406 | |

UBE2U
Genomic Map