siRNA Details
| Gene Symbol: LINC00337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103534.1 | 5 | 1104 | |||
| GenBank mRNA | BC030279.1 | - | 2 | 735 | 5141008 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00337| Gene Symbol: LINC00337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103534.1 | 5 | 1104 | |||
| GenBank mRNA | BC030279.1 | - | 2 | 735 | 5141008 | |