siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207396.2 | NP_997279.2 | 10 | 1068 | ||
| XM_011541439.2 | XP_011539741.1 | 9 | 1589 | |||
| XM_017001259.1 | XP_016856748.1 | 10 | 1022 | |||
| GenBank mRNA | AK056658.1 | - | 5 | 433 | ||
| AK294223.1 | - | 5 | 383 | |||
| BC034459.1 | - | 6 | 903 | |||
| BC128237.1 | - | 4 | 277 | |||
| DA190055.1 | - | 4 | 231 | |||

RNF207
Genomic Map