siRNA Details
| Gene Symbol: CCDC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152492.2 | NP_689705.2 | 4 | 675 | ||
| XM_011540780.1 | XP_011539082.1 | 4 | 678 | |||
| XM_017000402.1 | XP_016855891.1 | 4 | 678 | |||
| XM_017000403.1 | XP_016855892.1 | 4 | 678 | |||
| XM_017000404.1 | XP_016855893.1 | 4 | 678 | |||
| XM_017000405.1 | XP_016855894.1 | 4 | 678 | |||
| XM_017000406.1 | XP_016855895.1 | 5 | 492 | |||
| GenBank mRNA | AK057387.1 | - | 4 | 678 | ||
| BC112108.1 | - | 4 | 676 | 8327576 | ||
| BC112112.1 | - | 4 | 675 | 8327580 | ||

CCDC27
Genomic Map