siRNA Details
| Gene Symbol: CNIH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322305.1 | NP_001309234.1 | 1 | 713 | ||
| NM_152495.1 | NP_689708.1 | 1 | 713 | |||
| NR_136287.1 | 1 | 713 | ||||
| GenBank mRNA | AK096312.1 | - | 1 | 713 | ||
| BC022780.1 | - | 1 | 201 | 3894832 | ||

CNIH3
Genomic Map