siRNA Details
| Gene Symbol: EXOC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175876.4 | NP_787072.2 | 1 | 833 | ||
| GenBank mRNA | AK096460.1 | - | 1 | 813 | ||
| BC035763.1 | - | 1 | 779 | 5552837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXOC8| Gene Symbol: EXOC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175876.4 | NP_787072.2 | 1 | 833 | ||
| GenBank mRNA | AK096460.1 | - | 1 | 813 | ||
| BC035763.1 | - | 1 | 779 | 5552837 | ||