siRNA Details
| Gene Symbol: SZT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015284.3 | NP_056099.3 | 2 | 127 | ||
| XM_005270686.3 | XP_005270743.1 | 3 | 188 | |||
| XM_006710501.3 | XP_006710564.1 | 2 | 75 | |||
| XM_011541106.2 | XP_011539408.1 | 2 | 226 | |||
| XM_017000819.1 | XP_016856308.1 | 3 | 188 | |||
| XM_017000820.1 | XP_016856309.1 | 3 | 188 | |||
| GenBank mRNA | AK091821.1 | - | 2 | 48 | ||
| BC017576.1 | - | 2 | 131 | |||
| BC051343.1 | - | 2 | 61 | |||

SZT2
Genomic Map