siRNA Details
| Gene Symbol: FAM71A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153606.3 | NP_705834.2 | 1 | 2036 | ||
| GenBank mRNA | AK057358.1 | - | 1 | 1649 | ||
| AK301981.1 | - | 2 | 1282 | |||
| AK315021.1 | - | 1 | 1660 | |||
| BC035007.1 | - | 1 | 2011 | 5267155 | ||
| DB509538.1 | - | 1 | 142 | |||

FAM71A
Genomic Map