siRNA Details
| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008901.1 | NP_001008901.1 | 1 | 359 | ||
| NM_176791.3 | NP_789761.1 | 1 | 359 | |||
| XM_005260298.4 | XP_005260355.1 | 1 | 492 | |||
| GenBank mRNA | AA813527.1 | - | 2 | 402 | ||
| AK058060.1 | - | 1 | 355 | |||
| BC040049.1 | - | 1 | 284 | 5744556 | ||
| BQ428678.1 | - | 1 | 363 | |||
| BX355734.2 | - | 1 | 279 | |||

GTSF1L
Genomic Map