siRNA Details
| Gene Symbol: C20orf196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303477.1 | NP_001290406.1 | 4 | 503 | ||
| NM_001303478.1 | NP_001290407.1 | 4 | 482 | |||
| NM_152504.3 | NP_689717.2 | 3 | 412 | |||
| XM_006723549.3 | XP_006723612.1 | 3 | 343 | |||
| XM_011529177.2 | XP_011527479.1 | 3 | 399 | |||
| XM_017027684.1 | XP_016883173.1 | 4 | 514 | |||
| GenBank mRNA | BC035800.1 | - | 3 | 402 | 5734487 | |
| BU656896.1 | - | 4 | 456 | |||

C20orf196
Genomic Map