siRNA Details
| Gene Symbol: C20orf196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303477.1 | NP_001290406.1 | 4 | 783 | ||
| NM_001303478.1 | NP_001290407.1 | 4 | 762 | |||
| NM_152504.3 | NP_689717.2 | 3 | 692 | |||
| XM_006723549.3 | XP_006723612.1 | 3 | 623 | |||
| XM_011529177.2 | XP_011527479.1 | 3 | 679 | |||
| XM_017027684.1 | XP_016883173.1 | 4 | 794 | |||
| GenBank mRNA | AK057796.1 | - | 3, 4 | 527 | ||
| AK292708.1 | - | 3 | 678 | |||
| BC035800.1 | - | 3 | 682 | 5734487 | ||

C20orf196
Genomic Map