siRNA Details
| Gene Symbol: C2orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317992.1 | NP_001304921.1 | 5 | 644 | ||
| NM_144706.3 | NP_653307.1 | 4 | 548 | |||
| GenBank mRNA | AK314133.1 | - | 4 | 548 | ||
| BC021264.2 | - | 4 | 439 | 4542657 | ||
| BX364103.2 | - | 5 | 506 | |||
| DA396675.1 | - | 4 | 548 | |||

C2orf15
Genomic Map