siRNA Details
Gene Symbol: ZSWIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182521.2 | NP_872327.2 | 9 | 1178 | ||
GenBank mRNA | AK128006.1 | - | 9 | 1178 | ||
BC031094.1 | - | 9 | 1177 | 5298288 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZSWIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182521.2 | NP_872327.2 | 9 | 1178 | ||
GenBank mRNA | AK128006.1 | - | 9 | 1178 | ||
BC031094.1 | - | 9 | 1177 | 5298288 |