siRNA Details
| Gene Symbol: ZSWIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182521.2 | NP_872327.2 | 1, 2 | 195 | ||
| XM_011510712.1 | XP_011509014.1 | 1, 2 | 195 | |||
| GenBank mRNA | AK128006.1 | - | 1, 2 | 195 | ||
| BC031094.1 | - | 1, 2 | 194 | 5298288 | ||

ZSWIM2
Genomic Map