siRNA Details
| Gene Symbol: SLC16A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152527.4 | NP_689740.2 | 5 | 1887 | ||
| XM_005246353.3 | XP_005246410.1 | 4 | 1743 | |||
| XM_011510751.2 | XP_011509053.1 | 5 | 1658 | |||
| XM_017003480.1 | XP_016858969.1 | 5 | 1581 | |||
| GenBank mRNA | AK055317.1 | - | 2 | 337 | ||
| AK055356.1 | - | 3 | 1144 | |||
| AK292873.1 | - | 5 | 1619 | |||
| AL833470.1 | - | 2 | 386 | |||
| BC065524.1 | - | 5 | 1791 | 6058847 | ||
| BX648181.1 | - | 3 | 1055 | |||

SLC16A14
Genomic Map