siRNA Details
| Gene Symbol: SLC16A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152527.4 | NP_689740.2 | 3 | 830 | ||
| XM_011510751.2 | XP_011509053.1 | 3 | 601 | |||
| XM_011510752.1 | XP_011509054.1 | 3 | 830 | |||
| XM_017003480.1 | XP_016858969.1 | 3 | 524 | |||
| GenBank mRNA | AK123207.1 | - | 4 | 587 | ||
| AK292873.1 | - | 3 | 562 | |||
| BC034382.1 | - | 3 | 738 | |||
| BC065524.1 | - | 3 | 734 | 6058847 | ||

SLC16A14
Genomic Map